Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chlorpropham-d7 | 2140327-49-5 | 99.0% | 220.70 | C10H5D7ClNO2 | 1 MG
SDP

Catalog No. 5000302806
Encompass_Preferred

Chlorpropham-d7 is a deuterium-labeled isotopic standard of the herbicide chlorpropham intended for analytical and research applications. It is used as an internal standard or tracer in mass spectrometry and isotope-dilution studies to enable accurate quantification and method validation.

  • Deuterium-labeled isotopic standard for analytical use.
  • Molecular formula C10H5D7ClNO2 and molecular weight 220.70.
  • High purity: 99.0% (HPLC) as reported on the certificate of analysis.
  • Intended for use as an internal standard or tracer in mass spectrometry and quantitative analysis.
  • Storage: powder -20°C (stable for 3 years); in solvent -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-003-02806 Supplier Medchemexpress LLC Supplier No. HYW014240S11MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorpropham-d7 is a deuterium-labeled isotopic standard of the herbicide chlorpropham intended for analytical and research applications. It is used as an internal standard or tracer in mass spectrometry and isotope-dilution studies to enable accurate quantification and method validation.

  • Deuterium-labeled isotopic standard for analytical use.
  • Molecular formula C10H5D7ClNO2 and molecular weight 220.70.
  • High purity: 99.0% (HPLC) as reported on the certificate of analysis.
  • Intended for use as an internal standard or tracer in mass spectrometry and quantitative analysis.
  • Storage: powder -20°C (stable for 3 years); in solvent -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.