Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chlorpropham-d7 | 2140327-49-5 | 99.0% | 220.70 g/mol | C10H5D7ClNO2 | 5 MG
SDP

Catalog No. 5000302807
Encompass_Preferred

Chlorpropham-d7 is the deuterium-labeled analogue of the carbamate herbicide chlorpropham, supplied as an isotope-labeled analytical standard for mass spectrometry, metabolic tracing, and method development. The material is provided as a stable powder with specified isotopic enrichment and recommended storage conditions.

  • Deuterium-labeled isotope for tracer and quantitative studies.
  • Molecular formula C10H5D7ClNO2; molecular weight 220.70 g/mol.
  • High HPLC purity (99.0%) and isotopic enrichment (~99.3%).
  • Stable powder storage: -20°C; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Intended for research use only.

Catalog No. 50-003-02807 Supplier Medchemexpress LLC Supplier No. HYW014240S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorpropham-d7 is the deuterium-labeled analogue of the carbamate herbicide chlorpropham, supplied as an isotope-labeled analytical standard for mass spectrometry, metabolic tracing, and method development. The material is provided as a stable powder with specified isotopic enrichment and recommended storage conditions.

  • Deuterium-labeled isotope for tracer and quantitative studies.
  • Molecular formula C10H5D7ClNO2; molecular weight 220.70 g/mol.
  • High HPLC purity (99.0%) and isotopic enrichment (~99.3%).
  • Stable powder storage: -20°C; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.