Learn More
Medchemexpress LLC HY-B0274 50mg Medchemexpress, Chlorprothixene CAS:113-59-7 Purity:>98%
SDP
Catalog No. 502082440
Encompass_Preferred
Medchemexpress, HY-B0274 50mg Chlorprothixene CAS:113-59-7 Chlorprothixene is a dopamine and histamine receptors antagonist with Kis of 18 nM, 2.96 nM, 4.56 nM, 9 nM and 3.75 nM for hD1, hD2, hD3, hD5 and hH1 receptors, respectively. Antipsychotic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2440 Supplier Medchemexpress LLC Supplier No. HYB027450MG
Medchemexpress, HY-B0274 50mg Chlorprothixene CAS:113-59-7 Chlorprothixene is a dopamine and histamine receptors antagonist with Kis of 18 nM, 2.96 nM, 4.56 nM, 9 nM and 3.75 nM for hD1, hD2, hD3, hD5 and hH1 receptors, respectively. Antipsychotic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.