Learn More
Selleck Chemical LLC Chlorprothixene hydrochloride S5399-25mg
SDP
Catalog No. 503109513
Encompass_Preferred
Chlorprothixene HCl (Minithixen hydrochloride Truxal hydrochloride NSC 169899 NSC 56379 NSC 78193) is the hydrochloride salt form of chlorprothixene a typical antipsychotic drug of the thioxanthene (tricyclic) class It exerts strong blocking effects by blocking the 5-HT2 D1 D2 D3 histamine H1 muscarinic and alpha1 adrenergic receptors
Catalog No. 50-310-9513 Supplier Selleck Chemical LLC Supplier No. S539925MG
Chlorprothixene HCl (Minithixen hydrochloride Truxal hydrochloride NSC 169899 NSC 56379 NSC 78193) is the hydrochloride salt form of chlorprothixene a typical antipsychotic drug of the thioxanthene (tricyclic) class It exerts strong blocking effects by blocking the 5-HT2 D1 D2 D3 histamine H1 muscarinic and alpha1 adrenergic receptors