Learn More
Apexbio Technology LLC Chlorprothixene (hydrochloride) 6469-93-8 10g
SDP
Catalog No. 5000091475
Encompass_Preferred
Chlorprothixene hydrochloride (CAS 6469-93-8) is a small-molecule antagonist targeting dopamine (D1 D2 D3 D5) and histamine H1 receptors It modulates neurotransmission by blocking these receptors thereby regulating dopaminergic and histaminergic pathways Chlorprothixene hydrochloride exerts its biological activity primarily through receptor antagonism In in vitro studies it demonstrates nanomolar affinity with Ki values of 18 nM (D1) 2 96 nM (D2) 4 56 nM (D3) 9 nM (D5) and 3 75 nM (H1) Additionally it reduces SARS-CoV viral replication (IC50 13 0 18 5 M depending on strain) Based on these pharmacological properties chlorprothixene hydrochloride holds research potential in antipsychotic therapy antiviral investigation and modulation of inflammation in cystic fibrosis models
Catalog No. 50-000-91475 Supplier Apexbio Technology LLC Supplier No. C350910000
Chlorprothixene hydrochloride (CAS 6469-93-8) is a small-molecule antagonist targeting dopamine (D1 D2 D3 D5) and histamine H1 receptors It modulates neurotransmission by blocking these receptors thereby regulating dopaminergic and histaminergic pathways Chlorprothixene hydrochloride exerts its biological activity primarily through receptor antagonism In in vitro studies it demonstrates nanomolar affinity with Ki values of 18 nM (D1) 2 96 nM (D2) 4 56 nM (D3) 9 nM (D5) and 3 75 nM (H1) Additionally it reduces SARS-CoV viral replication (IC50 13 0 18 5 M depending on strain) Based on these pharmacological properties chlorprothixene hydrochloride holds research potential in antipsychotic therapy antiviral investigation and modulation of inflammation in cystic fibrosis models