Learn More
Medchemexpress LLC Chlorprothixene (hydrochloride) | 6469-93-8 | 99.9% | 200 MG
SDP
Catalog No. 5000408729
Encompass_Preferred
Chlorprothixene is a dopamine and histamine receptor antagonist known for its antipsychotic activity. It exhibits high affinity for various human dopamine and histamine receptors.
  • Antagonizes human D1, D2, D3, D5, and H1 receptors with low nanomolar Ki values.
  • Inhibits 4-(4-(dimethylamino)styryl)-N-methylpyridinium uptake at human OCT1 in HEK293 cells.
  • Antagonizes H1 receptor activity in human HeLa cells.
  • Binds to 5-HT7, 5-HT6, and 5-HT2 receptors with high pKi values.
Catalog No. 50-004-08729 Supplier Medchemexpress LLC Supplier No. HYB0274200MG
Chlorprothixene is a dopamine and histamine receptor antagonist known for its antipsychotic activity. It exhibits high affinity for various human dopamine and histamine receptors.
  • Antagonizes human D1, D2, D3, D5, and H1 receptors with low nanomolar Ki values.
  • Inhibits 4-(4-(dimethylamino)styryl)-N-methylpyridinium uptake at human OCT1 in HEK293 cells.
  • Antagonizes H1 receptor activity in human HeLa cells.
  • Binds to 5-HT7, 5-HT6, and 5-HT2 receptors with high pKi values.