Learn More
Medchemexpress LLC Chlorpyrifos | 2921-88-2 | 99.9% | 100 MG
SDP
Catalog No. 5000408192
Encompass_Preferred
Chlorpyrifos is a neurotoxic insecticide belonging to the class of thionite esters. It acts as an AChE inhibitor, impacting neurological function in insects, humans, and other animals. Chlorpyrifos interferes with cell replication and differentiation, ultimately altering synaptic transmission in neurons.
  • Acts as an AChE inhibitor
  • Interferes with cell replication and differentiation
  • Alters synaptic transmission in neurons
  • Neurotoxic insecticide
Catalog No. 50-004-08192 Supplier Medchemexpress LLC Supplier No. HYB0815100MG
Chlorpyrifos is a neurotoxic insecticide belonging to the class of thionite esters. It acts as an AChE inhibitor, impacting neurological function in insects, humans, and other animals. Chlorpyrifos interferes with cell replication and differentiation, ultimately altering synaptic transmission in neurons.
  • Acts as an AChE inhibitor
  • Interferes with cell replication and differentiation
  • Alters synaptic transmission in neurons
  • Neurotoxic insecticide