Learn More
Cayman Chemical Chlorpyrifos, Purity: ≥98%, CAS: 2921-88-2, 50 mg
SDP
Catalog No. 502994249
Encompass_Preferred
An organophosphate insecticide; lethal to A. melinus, G. ashmeadi, E. eremicus, and E. formosa adults (LC50s = 0.8, 6, 12, and 17 ng/ml, respectively); induces mortality in O. insidiosus adults when applied to corn, sorghum, and alfalfa plants; toxic to mice (LD50 = 155 mg/kg); Postnatal day 11 to 14 exposure decreases nest building and defense behaviors in adult female mice at 3 mg/kg
Catalog No. 50-299-4249 Supplier Cayman Chemical Supplier No. 2141250 MG
An organophosphate insecticide; lethal to A. melinus, G. ashmeadi, E. eremicus, and E. formosa adults (LC50s = 0.8, 6, 12, and 17 ng/ml, respectively); induces mortality in O. insidiosus adults when applied to corn, sorghum, and alfalfa plants; toxic to mice (LD50 = 155 mg/kg); Postnatal day 11 to 14 exposure decreases nest building and defense behaviors in adult female mice at 3 mg/kg