Learn More
Medchemexpress LLC HY-B0815 50mg Medchemexpress, Chlorpyrifos CAS:2921-88-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB081550MG
Encompass_Preferred
Medchemexpress, HY-B0815 50mg Chlorpyrifos CAS:2921-88-2 Chlorpyrifos is an organophosphate insecticide that is classified as a phosphorothionate. The oxon metabolite of Chlorpyrifos is an inhibitor of acetylcholinesterase (AChE), affecting neurological function in insects, humans, and other animals. The Chlorpyrifos oxon (CPO) metabolite is hydrolyzed by the plasma enzyme paraoxonase 1 (PON1), and susceptibility to neurotoxicity associated with CPO exposure is mitigated by PON1 overexpression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1128