Learn More
Medchemexpress LLC Chlorpyrifos-d10 | 285138-81-0 | 99.5% | 1 MG
SDP
Catalog No. 5000409382
Encompass_Preferred
Chlorpyrifos-d10 is the deuterium labeled Chlorpyrifos. Chlorpyrifos is an organophosphate insecticide classified as a phosphorothionate. Its oxon metabolite inhibits acetylcholinesterase (AChE), affecting neurological function in insects, humans, and other animals. It can be used as a tracer or an internal standard for quantitative analysis.
  • Deuterium labeled compound
  • Organophosphate insecticide
  • Oxon metabolite inhibits acetylcholinesterase (AChE)
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-09382 Supplier Medchemexpress LLC Supplier No. HYB0815S1MG
Chlorpyrifos-d10 is the deuterium labeled Chlorpyrifos. Chlorpyrifos is an organophosphate insecticide classified as a phosphorothionate. Its oxon metabolite inhibits acetylcholinesterase (AChE), affecting neurological function in insects, humans, and other animals. It can be used as a tracer or an internal standard for quantitative analysis.
  • Deuterium labeled compound
  • Organophosphate insecticide
  • Oxon metabolite inhibits acetylcholinesterase (AChE)
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS