Learn More
Medchemexpress LLC Chlorpyrifos-d10 | 285138-81-0 | MFCD01321400 | 99.5% | 360.65 | C9HD10Cl3NO3PS | 10 MG
SDP
Catalog No. 5000297010
Encompass_Preferred
Chlorpyrifos-d10 is the deuterium-labeled analog of the organophosphate insecticide chlorpyrifos, provided as a high-purity reference standard for analytical and toxicology research. It is supplied as a powder for use as an internal standard in GC- or LC-MS workflows and environmental residue testing.
  • Deuterium-labeled internal standard for analytical quantification.
  • High purity (~99.5%) suitable for trace analysis.
  • Powder form for easy dissolution and spiking into samples.
  • Available in mg-scale packs, e.g., 10 mg, for laboratory use.
  • Stable under recommended frozen storage for extended shelf life.
Catalog No. 50-002-97010 Supplier Medchemexpress LLC Supplier No. HYB0815S10MG
Chlorpyrifos-d10 is the deuterium-labeled analog of the organophosphate insecticide chlorpyrifos, provided as a high-purity reference standard for analytical and toxicology research. It is supplied as a powder for use as an internal standard in GC- or LC-MS workflows and environmental residue testing.
  • Deuterium-labeled internal standard for analytical quantification.
  • High purity (~99.5%) suitable for trace analysis.
  • Powder form for easy dissolution and spiking into samples.
  • Available in mg-scale packs, e.g., 10 mg, for laboratory use.
  • Stable under recommended frozen storage for extended shelf life.