Learn More
Crescent Chemical Co Inc CHLORPYRIFOS-METHYL 250 MG
SDP
Catalog No. 5073060
Encompass_Preferred
Chlorpyrifos-methyl Insecticide (WS) CAS 5598-13-0 Molecular Weight - 322.5331 Storage - -18°C Catalog Num - DRE-C11601000 pesticide
Catalog No. 50-730-60 Supplier Crescent Chemical Co Inc Supplier No. PSC11601000
Chlorpyrifos-methyl Insecticide (WS) CAS 5598-13-0 Molecular Weight - 322.5331 Storage - -18°C Catalog Num - DRE-C11601000 pesticide