Learn More
Crescent Chemical Co Inc CHLORPYRIFOS SOLN 1 ML
SDP
Catalog No. 5079997
Encompass_Preferred
Chlorpyrifos 100 µg/mL in Acetonitrile Insecticide (WS solution) CAS 2921-88-2 Molecular Weight - 350.5863 Storage - RT Catalog Num - DRE-XA11600000AL pesticide
Catalog No. 50-799-97 Supplier Crescent Chemical Co Inc Supplier No. PSXA11600000AL
Chlorpyrifos 100 µg/mL in Acetonitrile Insecticide (WS solution) CAS 2921-88-2 Molecular Weight - 350.5863 Storage - RT Catalog Num - DRE-XA11600000AL pesticide