Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Technologies, Inc. Organic Standard, Chlorpyriphos, Neat, 100 mg

Catalog No. USPST480
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PST-480 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PST-480 Supplier Agilent Technologies Supplier No. PST480
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2921-88-2
Molecular Formula C9H11Cl3NO3PS
Molecular Weight (g/mol) 350.575
InChI Key SBPBAQFWLVIOKP-UHFFFAOYSA-N
PubChem CID 2730
ChEBI CHEBI:34631
IUPAC Name diethoxy-sulfanylidene-(3,5,6-trichloropyridin-2-yl)oxy-$l^{5}-phosphane
SMILES CCOP(=S)(OCC)OC1=NC(=C(C=C1Cl)Cl)Cl

Specifications

CAS Max % 100.0
Assay Percent Range >95 % min.
Solvent or Matrix Neat
Quantity 100 mg
Chemical Name or Material Chlorpyriphos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.