Learn More
Apexbio Technology LLC Chlorquinaldol 72-80-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061988
Encompass_Preferred
Chlorquinaldol is a synthetic antimicrobial agent exhibiting antifungal and antibacterial properties commonly utilized in biomedical research for evaluating local antiseptic mechanisms It functions primarily through disrupting microbial membrane integrity and interfering with essential enzymatic pathways ultimately limiting microbial proliferation Chlorquinaldol is frequently employed in vitro for susceptibility testing and characterizing antimicrobial activity against various fungal and bacterial strains In experimental assays its antifungal activity is evaluated through measurement of inhibitory concentrations with reported IC50 values typically ranging between 10-50 g/mL depending on specific microorganism types and assay conditions This compound serves as a reference tool in microbiological studies addressing drug susceptibility resistance evaluation and antiseptic compound benchmarking
Catalog No. 50-000-61988 Supplier Apexbio Technology LLC Supplier No. B16985.1
Chlorquinaldol is a synthetic antimicrobial agent exhibiting antifungal and antibacterial properties commonly utilized in biomedical research for evaluating local antiseptic mechanisms It functions primarily through disrupting microbial membrane integrity and interfering with essential enzymatic pathways ultimately limiting microbial proliferation Chlorquinaldol is frequently employed in vitro for susceptibility testing and characterizing antimicrobial activity against various fungal and bacterial strains In experimental assays its antifungal activity is evaluated through measurement of inhibitory concentrations with reported IC50 values typically ranging between 10-50 g/mL depending on specific microorganism types and assay conditions This compound serves as a reference tool in microbiological studies addressing drug susceptibility resistance evaluation and antiseptic compound benchmarking