Learn More
Medchemexpress LLC Chlorquinaldol | 72-80-0 | 98.7% | 500 MG
SDP
Catalog No. 5000413043
Encompass_Preferred
Chlorquinaldol (Chloquinan) is an antibacterial agent with potential use in topical skin conditions and vaginal infections. It is also a β-catenin/TCF4 inhibitor, demonstrating anti-proliferation, anti-migration, and apoptosis-inducing activity in cancer cells.
  • Antibacterial agent for topical skin and vaginal infections.
  • Acts as a β-catenin/TCF4 inhibitor.
  • Demonstrates anti-proliferation and anti-migration activity.
  • Induces apoptosis in cancer cells.
  • Inhibits Wnt/β-catenin signaling in colorectal cancer cells.
  • Reduces β-catenin acetylation, affecting interaction with TCF4.
  • Inhibits stemness of colorectal cancer cells.
Catalog No. 50-004-13043 Supplier Medchemexpress LLC Supplier No. HYB1360500MG
Chlorquinaldol (Chloquinan) is an antibacterial agent with potential use in topical skin conditions and vaginal infections. It is also a β-catenin/TCF4 inhibitor, demonstrating anti-proliferation, anti-migration, and apoptosis-inducing activity in cancer cells.
  • Antibacterial agent for topical skin and vaginal infections.
  • Acts as a β-catenin/TCF4 inhibitor.
  • Demonstrates anti-proliferation and anti-migration activity.
  • Induces apoptosis in cancer cells.
  • Inhibits Wnt/β-catenin signaling in colorectal cancer cells.
  • Reduces β-catenin acetylation, affecting interaction with TCF4.
  • Inhibits stemness of colorectal cancer cells.