Learn More
Bioworld Chlorsulfuron, 100 mg
Catalog No. 50255082
Encompass_Preferred
ChlorsulfuronSynonym: 1-(2-Chlorophenylsulfonyl)-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)-ureaAffects the biosynthesis of branched chain amino acids. It has also been applied as a successful selective agent in the transformation of tobacco, maize, and sugarbeetsAppearance: white crystaline solidMelting Point: 174-178 °CSoluble in water; Slightly soluble in methylene chloride Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment
Catalog No. 50-255-082 Supplier Bioworld Supplier No. 306320011
ChlorsulfuronSynonym: 1-(2-Chlorophenylsulfonyl)-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)-ureaAffects the biosynthesis of branched chain amino acids. It has also been applied as a successful selective agent in the transformation of tobacco, maize, and sugarbeetsAppearance: white crystaline solidMelting Point: 174-178 °CSoluble in water; Slightly soluble in methylene chloride Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment