Learn More
Medchemexpress LLC Chlortetracycline-13C,d3 hydrochloride | 519.35 g/mol | C21 13CH21D3Cl2N2O8 | 500 UG
SDP
Catalog No. 5000284369
Encompass_Preferred
Chlortetracycline-13C,d3 hydrochloride is the 13C- and deuterium-labeled form of chlortetracycline hydrochloride supplied as an analytical reference standard for use in mass spectrometry, chromatographic method development, and tracer studies. It is provided in a microgram pack suitable for analytical laboratories.
  • Isotope-labeled standard for mass spectrometry and tracer studies.
  • Labeled with 13C and d3 for clear mass-shift differentiation.
  • Supplied as the hydrochloride salt for improved stability.
  • Molecular weight 519.35 g/mol.
  • Packaged in a 500 UG quantity suitable for analytical use.
Catalog No. 50-002-84369 Supplier Medchemexpress LLC Supplier No. HYW653977500UG
Chlortetracycline-13C,d3 hydrochloride is the 13C- and deuterium-labeled form of chlortetracycline hydrochloride supplied as an analytical reference standard for use in mass spectrometry, chromatographic method development, and tracer studies. It is provided in a microgram pack suitable for analytical laboratories.
  • Isotope-labeled standard for mass spectrometry and tracer studies.
  • Labeled with 13C and d3 for clear mass-shift differentiation.
  • Supplied as the hydrochloride salt for improved stability.
  • Molecular weight 519.35 g/mol.
  • Packaged in a 500 UG quantity suitable for analytical use.