Learn More
Apexbio Technology LLC Chlortetracycline HCl 64-72-2 250mg
SDP
Catalog No. 502240578
Encompass_Preferred
Chlortetracycline HCl (64-72-2) is a small-molecule inhibitor targeting the bacterial 30S ribosomal subunit It is designed to inhibit protein synthesis by interfering with aminoacyl-tRNA binding and preventing elongation of the polypeptide chain Chlortetracycline HCl exerts its biological activity primarily through reversible binding to the bacterial 30S ribosomal subunit resulting in inhibition of bacterial protein synthesis Based on these pharmacological properties Chlortetracycline HCl holds research potential in controlling bacterial contamination in cell culture systems studying bacterial resistance mechanisms evaluating antibiotic susceptibility and investigating microbial protein biosynthesis pathways in vitro
This item is not returnable. View return policy
Catalog No. 50-224-0578 Supplier Apexbio Technology LLC Supplier No. B1699250
Chlortetracycline HCl (64-72-2) is a small-molecule inhibitor targeting the bacterial 30S ribosomal subunit It is designed to inhibit protein synthesis by interfering with aminoacyl-tRNA binding and preventing elongation of the polypeptide chain Chlortetracycline HCl exerts its biological activity primarily through reversible binding to the bacterial 30S ribosomal subunit resulting in inhibition of bacterial protein synthesis Based on these pharmacological properties Chlortetracycline HCl holds research potential in controlling bacterial contamination in cell culture systems studying bacterial resistance mechanisms evaluating antibiotic susceptibility and investigating microbial protein biosynthesis pathways in vitro