Learn More
Bioworld Chlortetracycline HCl, 25 g
Catalog No. 50255719
Encompass_Preferred
Chlortetracycline, HCl Bacteriostatic antibiotic active against Gram-positive and Gram-negative bacteria.Protect from light.A specific and potent calcium ionophore antibioticFor in vitro lab use only. Not for human use or consumption.Appearance: SolidBoiling Point : 828.9 °CMelting Point: 210-215 °C (lit.)(dec.)RTECS Number: QI7800000
Catalog No. 50-255-719 Supplier Bioworld Supplier No. 403100201
Chlortetracycline, HCl Bacteriostatic antibiotic active against Gram-positive and Gram-negative bacteria.Protect from light.A specific and potent calcium ionophore antibioticFor in vitro lab use only. Not for human use or consumption.Appearance: SolidBoiling Point : 828.9 °CMelting Point: 210-215 °C (lit.)(dec.)RTECS Number: QI7800000