Learn More
Enzo Life Sciences Chlortetracycline . hydrochloride (5g). CAS: 64-72-2
SDP
Supplier:  Enzo Life Sciences ALX350238G005
Encompass_Preferred
Antibiotic. Inhibits protein synthesis in prokaryotes and eukaryotes. Inhibits binding of aminoacyl-tRNA to ribosomes. Used as a fluorescent Ca2+ probe. Alternative name: Aureomycin . HCI. Purity: ≥85% (UHPLC). Solubility: Soluble in 1M NaOH (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8793