Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlortetracycline HCl, USP Grade, G-Biosciences
SCI_ED

Catalog No. S720061
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S720061 100 g
S955274 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S720061 Supplier G-Biosciences Supplier No. RC969
Add to Cart
Add to Cart

Chlortetracycline, isolated from Streptomyces aureofaciens, was the first tetracycline antibiotic discovered. It has shown the most efficacy against gram-positive organisms, but is also effective on a variety of gram-negative bacteria.

Resistance to chlortetracycline arises from a loss of cell wall permeability.

Chemical Identifiers

Molecular Weight (g/mol) 515.34 g/mol

Specifications

Boiling Point 210 to 215°C
Quantity 100 g
Percent Purity >94.5%
Grade USP
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.