Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Chlortetracycline hydrochloride, can be used as secondary standard

Catalog No. AC268245000
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC268245000 500 mg Glass Bottle
AC268240025 2.5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC268245000 Supplier Thermo Scientific Chemicals Supplier No. 268245000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chlortetracycline hydrochloride, CAS # 64-72-2, is a broad-spectrum antibiotic of the tetracycline family

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Chlortetracycline hydrochloride is a broad-spectrum antibiotic of the tetracycline family
  • Receives its name from a 7-chloro substitution on one of the rings of its parent structure, tetracycline
  • A bacteriostatic that inhibits the 30S ribosomal subunit and prevents peptide formation
  • Can work as an antiprotozoal and antifungal

Applications

  • Chlortetracycline is suitable for use in biochemistry, microbiology, in vivo research, and cell culture
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-72-2
Molecular Formula C22H24Cl2N2O8
Molecular Weight (g/mol) 515.34
InChI Key QYAPHLRPFNSDNH-CIVPRPTRSA-N
Synonym CTC
PubChem CID 66577600
IUPAC Name (4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl

Specifications

Type Chlortetracycline Hydrochloride
Physical Form Fine Crystalline Powder
Melting Point 220.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation − 242.50 (20.00°C c=0.5,water)
Color Yellow
Quantity 500 mg
Solubility Information Solubility in water: 8.6g/L (28°C). Other solubilities: practically insoluble in acetone, ether,, chloroform, dioxane and propylene glycol, 17.4g/L methanol (28°C), 1.7g/L ethanol (28°C), soluble in solutions of alkali hydroxides and, carbonate
Formula Weight 515.33
Percent Purity ≥80 wt% (as is, calculated) (HPLC)
Chemical Name or Material Chlortetracycline hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.