Learn More
Medchemexpress LLC HY-15833 50mg , Chlorthalidone CAS:77-36-1 Purity:98%
SDP
Catalog No. 501493788
Encompass_Preferred
Medchemexpress, HY-15833 50mg Chlorthalidone CAS:77-36-1 Formula:C14H11ClN2O4S Purity:98% Chlorthalidone is a thiazide-like diuretic used to treat hypertension. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3788 Supplier Medchemexpress LLC Supplier No. HY1583350MG
Medchemexpress, HY-15833 50mg Chlorthalidone CAS:77-36-1 Formula:C14H11ClN2O4S Purity:98% Chlorthalidone is a thiazide-like diuretic used to treat hypertension. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.