Learn More
Cayman Chemical Chlorzoxazon-d3 500ug
SDP
Catalog No. 502948066
Encompass_Preferred
An internal standard for the quantification of chlorzoxazone by GC- or LC-MS
Catalog No. 50-294-8066 Supplier Cayman Chemical Supplier No. 34237500 UG
An internal standard for the quantification of chlorzoxazone by GC- or LC-MS