Learn More
Apexbio Technology LLC Chlorzoxazone 95-25-0 5g
SDP
Catalog No. 502240777
Encompass_Preferred
Chlorzoxazone (CAS 95-25-0) is a small molecule that functions as a centrally acting skeletal muscle relaxant It exerts its pharmacological effects by modulating synaptic transmission within the spinal cord resulting in reduced muscle spasticity In biomedical research chlorzoxazone is frequently employed as a selective probe substrate for cytochrome P450 2E1 (CYP2E1) in human liver microsomes facilitating the assessment of hepatic CYP2E1 activity and drug metabolism This compound is valuable for investigating drug-drug interactions and metabolic profiling involving CYP2E1
This item is not returnable. View return policy
Catalog No. 50-224-0777 Supplier Apexbio Technology LLC Supplier No. B17015000
Chlorzoxazone (CAS 95-25-0) is a small molecule that functions as a centrally acting skeletal muscle relaxant It exerts its pharmacological effects by modulating synaptic transmission within the spinal cord resulting in reduced muscle spasticity In biomedical research chlorzoxazone is frequently employed as a selective probe substrate for cytochrome P450 2E1 (CYP2E1) in human liver microsomes facilitating the assessment of hepatic CYP2E1 activity and drug metabolism This compound is valuable for investigating drug-drug interactions and metabolic profiling involving CYP2E1