Learn More
Medchemexpress LLC Chlorzoxazone-d3 | 1185173-60-7 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1462S5MG
Encompass_Preferred
Chlorzoxazone-d3 is a deuterium-labeled form of Chlorzoxazone. Chlorzoxazone is a centrally acting muscle relaxant used to alleviate muscle spasms and associated pain or discomfort. It functions by depressing reflexes in the spinal cord. Chlorzoxazone is also utilized as a marker substrate in in vitro and in vivo studies to quantify the activity of cytochrome P450 2E1 (CYP2E1) in humans.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11889