Learn More
Chiral Technologies, Inc. CHLPAK IG-3, 150X2.1MM, 3UM CO
Catalog No. NC2201637
Encompass
CHIRALPAK IG-3, 150x2.1mm, 3um particle size HPLC column
This item is not returnable. View return policy
Catalog No. NC2201637 Supplier Chiral Technologies, Inc. Supplier No. 87594
CHIRALPAK IG-3, 150x2.1mm, 3um particle size HPLC column