Learn More
TARGETMOL CHEMICALS INC CHM-1 5MG
SDP
Catalog No. 502580936
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CHM-1 is an inducer of apoptosis and displays potent antitumor ability in human hepatocellular carcinoma by activation of Cdc2 kinase activity. CHM-1 inhibits tubulin polymerization in vitro and in vivo. purity: 100%
Catalog No. 50-258-0936 Supplier TARGETMOL CHEMICALS INC Supplier No. T226615MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CHM-1 is an inducer of apoptosis and displays potent antitumor ability in human hepatocellular carcinoma by activation of Cdc2 kinase activity. CHM-1 inhibits tubulin polymerization in vitro and in vivo. purity: 100%