Learn More
Aviva Systems Biology Corp CHM Antibody - N-terminal region (AVARP13011_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13011_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHM. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHM. Alternate names: DXS540, FLJ38564, GGTA, HSD-32, MGC102710, REP-1, TCDApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-62600