Learn More
Abcam CHM1, Antitumor agent, 50MG
Catalog No. 502706782
Encompass_Preferred
MW 283.25 Da, Purity >99%. Potent antitumor agent. Antiproliferative, anti-angiogenic and anti-apoptotic in a dose dependent manner. Reduces cell viability (IC₅₀ = 0.13 - 8.2 μM) and tumor invasion. Active in vitro and in vivo. Increases expression of DR5, TRIAL, p53. Reduces tumor polymerisation and caspase activation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6782 Supplier Abcam Supplier No. AB14122550MG
MW 283.25 Da, Purity >99%. Potent antitumor agent. Antiproliferative, anti-angiogenic and anti-apoptotic in a dose dependent manner. Reduces cell viability (IC₅₀ = 0.13 - 8.2 μM) and tumor invasion. Active in vitro and in vivo. Increases expression of DR5, TRIAL, p53. Reduces tumor polymerisation and caspase activation.
The product is subject to the following: Abcam Restricted Use Statement