Learn More
Medchemexpress LLC HY-101267 100mg Medchemexpress, CHMFL-BMX-078 CAS:1808288-51-8 Purity:>98%
SDP
Catalog No. 501932079
Encompass_Preferred
Medchemexpress, HY-101267 100mg CHMFL-BMX-078 CAS:1808288-51-8 CHMFL-BMX-078 is a highly potent and selective type II irreversible BMX kinase inhibitor with an IC 50 of 11 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2079 Supplier Medchemexpress LLC Supplier No. HY101267100MG
Medchemexpress, HY-101267 100mg CHMFL-BMX-078 CAS:1808288-51-8 CHMFL-BMX-078 is a highly potent and selective type II irreversible BMX kinase inhibitor with an IC 50 of 11 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.