Learn More
Medchemexpress LLC CHMFL-FLT3-122 | 1839150-56-9 | 98.5% | 471.55 | 1 ML
SDP
Catalog No. 5000343616
Encompass_Preferred
CHMFL-FLT3-122 is a potent, selective, and orally active FLT3 kinase inhibitor with an IC50 of 40 nM. It demonstrates selectivity for FLT3 over BTK and c-KIT kinases. This compound induces apoptosis by arresting the cell cycle in the G0/G1 phase and has shown to significantly inhibit the proliferation of FLT3-ITD positive acute myeloid leukemia (AML) cancer cell lines in in vitro studies. In in vivo studies, it suppressed tumor progression without significantly affecting body weight.
  • Potent and selective FLT3 kinase inhibitor
  • Orally active
  • Induces apoptosis by arresting cell cycle in G0/G1 phase
  • Inhibits proliferation of FLT3-ITD positive AML cell lines
  • Inhibits FLT3 auto-phosphorylation, Stat5, ERK, AKT phosphorylation, and cMYC expression
  • Suppresses tumor progression in in vivo models
Catalog No. 50-003-43616 Supplier Medchemexpress LLC Supplier No. HY11029350MG
CHMFL-FLT3-122 is a potent, selective, and orally active FLT3 kinase inhibitor with an IC50 of 40 nM. It demonstrates selectivity for FLT3 over BTK and c-KIT kinases. This compound induces apoptosis by arresting the cell cycle in the G0/G1 phase and has shown to significantly inhibit the proliferation of FLT3-ITD positive acute myeloid leukemia (AML) cancer cell lines in in vitro studies. In in vivo studies, it suppressed tumor progression without significantly affecting body weight.
  • Potent and selective FLT3 kinase inhibitor
  • Orally active
  • Induces apoptosis by arresting cell cycle in G0/G1 phase
  • Inhibits proliferation of FLT3-ITD positive AML cell lines
  • Inhibits FLT3 auto-phosphorylation, Stat5, ERK, AKT phosphorylation, and cMYC expression
  • Suppresses tumor progression in in vivo models