Learn More
Medchemexpress LLC CHMFL-PI3KD-317 | 2244992-76-3 | 98.0% | 494.03 | 100 MG
SDP
Catalog No. 5000437542
Encompass_Preferred
CHMFL-PI3KD-317 is a highly potent, selective, and orally active PI3Kδ inhibitor with an IC50 of 6 nM. It demonstrates over 10-1500 fold selectivity compared to other class I, II, and III PIKK family isoforms. The compound inhibits PI3Kδ-mediated Akt T308 phosphorylation in Raji cells with an EC50 of 4.3 nM and exhibits antiproliferative effects on cancer cells.
  • Highly potent and selective PI3Kδ inhibitor
  • Orally active
  • Exhibits antiproliferative effects on cancer cells
  • Favorable oral bioavailability
  • Acceptable half-life
Catalog No. 50-004-37542 Supplier Medchemexpress LLC Supplier No. HY112608100MG
CHMFL-PI3KD-317 is a highly potent, selective, and orally active PI3Kδ inhibitor with an IC50 of 6 nM. It demonstrates over 10-1500 fold selectivity compared to other class I, II, and III PIKK family isoforms. The compound inhibits PI3Kδ-mediated Akt T308 phosphorylation in Raji cells with an EC50 of 4.3 nM and exhibits antiproliferative effects on cancer cells.
  • Highly potent and selective PI3Kδ inhibitor
  • Orally active
  • Exhibits antiproliferative effects on cancer cells
  • Favorable oral bioavailability
  • Acceptable half-life