Learn More
Medchemexpress LLC Chmfl-pi4k-127 | 2377604-81-2 | 99.9% | 402.85 g/mol | C18H15ClN4O3S | 10 MG
SDP
Catalog No. 5000287240
Encompass_Preferred
CHMFL-PI4K-127 is a potent, selective Plasmodium falciparum phosphatidylinositol 4-kinase (PfPI4K) inhibitor provided for research use. It exhibits sub-nanomolar enzymatic potency and cellular antimalarial activity, and is supplied at high chemical purity suitable for in vitro and in vivo studies.
  • Potent PfPI4K inhibition (IC50 ~0.9 nM).
  • Demonstrated cellular antimalarial activity (EC50 ~25.1 nM).
  • High chemical purity for reliable experimental results (>99.9%).
  • Available in research-scale quantities for assay development.
  • Soluble in DMSO for standard compound handling and dosing.
Catalog No. 50-002-87240 Supplier Medchemexpress LLC Supplier No. HY15059810MG
CHMFL-PI4K-127 is a potent, selective Plasmodium falciparum phosphatidylinositol 4-kinase (PfPI4K) inhibitor provided for research use. It exhibits sub-nanomolar enzymatic potency and cellular antimalarial activity, and is supplied at high chemical purity suitable for in vitro and in vivo studies.
  • Potent PfPI4K inhibition (IC50 ~0.9 nM).
  • Demonstrated cellular antimalarial activity (EC50 ~25.1 nM).
  • High chemical purity for reliable experimental results (>99.9%).
  • Available in research-scale quantities for assay development.
  • Soluble in DMSO for standard compound handling and dosing.