Learn More
Aviva Systems Biology Corp CHML Antibody - N-terminal region (AVARP13012_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13012_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHML. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHML. Alternate names: REP2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62601