Learn More
Aviva Systems Biology Corp CHMP1B Antibody - N-terminal region (ARP57408_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57408_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHMP1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHMP1B. Alternate names: C10orf2, C18-ORF2, C18orf2, CHMP1.5, Vps46-2, Vps46B, hVps46-2ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53989