Learn More
Aviva Systems Biology Corp CHMP4A Antibody - C-terminal region (ARP61976_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61976_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHMP4A. It was validated on Western Blot. . Alternate names: C14orf123, CHMP4, CHMP4B, FLJ61658, HSPC134, MGC142093, MGC142095, SNF7, SNF7-1, Shax2, VPS32A, SHAX2, VPS32-1ApplicationWB. Species reactivity: Cow, Human, Rabbit, Rat
Catalog No. 50-287-529