Learn More
Aviva Systems Biology Corp CHMP4B Antibody - middle region (ARP53339_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53339_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHMP4B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CHMP4B. Alternate names: C20orf178, CHMP4A, CTPP3, SNF7, SNF7-2, Shax1, dJ553F4.4, VPS32B, Vps32-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-828