Learn More
Aviva Systems Biology Corp CHMP6 Antibody - middle region (ARP65538_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65538_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHMP6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human CHMP6. Alternate names: FLJ11749, VPS20ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-288-515