Learn More
Aviva Systems Biology Corp CHN1 Antibody - C-terminal region (ARP51974_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51974_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHN1. It was validated on Western Blot. . Alternate names: ARHGAP2, CHN, DURS2, RHOGAP2, NCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-617