Learn More
Aviva Systems Biology Corp CHN2 Antibody - N-terminal region (ARP51863_P050) | 100uL
SDP
Catalog No. 5001460632
Encompass_Preferred
This is a rabbit polyclonal antibody against CHN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHN2. Alternate names: ARHGAP3, BCH, MGC138360, RHOGAP3, CHN2-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60632 Supplier Aviva Systems Biology Corp Supplier No. ARP51863_P050
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This is a rabbit polyclonal antibody against CHN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHN2. Alternate names: ARHGAP3, BCH, MGC138360, RHOGAP3, CHN2-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish