Learn More
ATCC CHO 1-15 500; Ovary; Chinese Hamster (Cricetulus griseus)
Supplier:  ATCC CRL9606
Encompass
CHO 1-15 [subscript 500] is an epithelial cell line that was isolated from the ovary of an adult female Chinese hamster. The line was produced by transfecting CHO cells with a plasmid (pETPFR, ATCC 40403) to produce a system in which a human tissue plasminogen activator is produced.
Catalog No. 50-238-3186