Learn More
Lonza Walkersville CHO 7 Lipids 2x contr. 500 mL
Catalog No. 5000496316
Encompass
CHO 7 Lipids can be ordered separately or is included in a ProCHO or PowerCHO Powder Kit This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96316 Supplier Lonza Walkersville Supplier No. BE13668F
CHO 7 Lipids can be ordered separately or is included in a ProCHO or PowerCHO Powder Kit This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS