Learn More
Cellular Engineering Technologies CHO Cell Culture Growth Media (450mL)
Supplier:  Cellular Engineering Technologies MR1015
Encompass_Preferred
Media is specifically designed for high-density growth of adherent and suspension Chinese Hamster Ovary (CHO) cell cultures. This media is transfection-compatible with available transfection reagents and maintains high cell viability at high cell densities. Media contains fetal bovine serum.Media is shipped with gel packs.
Catalog No. NC2753089