Learn More
Expression Systems CHO-ES (suspension CHO K1) cells, Frozen in ESF SFM, 20M cells/vial
SDP
Catalog No. NC1481663
Encompass_Preferred
Expression Systems offers CHO mammalian cell lines adapted to serum-free suspension culture in ESF SFM. The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
CHO-ES cells are derived from the CHO (Chinese Hamster Ovary) cell line. CHO-ES cells are adapted for suspension culture in ESF SFM and are available as a frozen vial.
• Capable of high levels of recombinant protein expression
• Good growth in adherent or suspension culture
• Quality and performance testing
Organism: Cricetulus griseus, hamster, Chinese
Tissue: Ovary
Cell Type: Epithelial
Biosafety Level 1: Biosafety classification is based on U.S. Public Health Service Guidelines, it is the responsibility of the customer to ensure that their facilities comply with biosafety regulations for their own country.
To download Instructions for Use and/or COA please visit: www.expressionsystems.com
Catalog No. NC1481663 Supplier Expression Systems Supplier No. 94008F
Expression Systems offers CHO mammalian cell lines adapted to serum-free suspension culture in ESF SFM. The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
CHO-ES cells are derived from the CHO (Chinese Hamster Ovary) cell line. CHO-ES cells are adapted for suspension culture in ESF SFM and are available as a frozen vial.
• Capable of high levels of recombinant protein expression
• Good growth in adherent or suspension culture
• Quality and performance testing
Organism: Cricetulus griseus, hamster, Chinese
Tissue: Ovary
Cell Type: Epithelial
Biosafety Level 1: Biosafety classification is based on U.S. Public Health Service Guidelines, it is the responsibility of the customer to ensure that their facilities comply with biosafety regulations for their own country.
To download Instructions for Use and/or COA please visit: www.expressionsystems.com