Learn More
Cygnus Technologies CHO DNA Amplification Kit, Residual DNA Quantification, qPCR Compatible, 1 kit
Catalog No. NC3799809
Encompass
The CHO DNA Amplification Kit is a reliable solution for precise quantification of residual host cell DNA in biopharmaceutical products derived from CHO cell lines. Utilizing a proprietary DNA extraction process, it effectively isolates HCDNA while minimizing interference, ensuring accurate results vital for process development, lot release, and quality control. Designed specifically for research, development, and manufacturing applications, it stands out with compatibility for existing quantitative PCR instruments.
Catalog No. NC3799809 Supplier Cygnus Technologies Supplier No. D555T
The CHO DNA Amplification Kit is a reliable solution for precise quantification of residual host cell DNA in biopharmaceutical products derived from CHO cell lines. Utilizing a proprietary DNA extraction process, it effectively isolates HCDNA while minimizing interference, ensuring accurate results vital for process development, lot release, and quality control. Designed specifically for research, development, and manufacturing applications, it stands out with compatibility for existing quantitative PCR instruments.