Learn More
Eurofins Discoverx PathHunter CHO-K1 DRD2S (Short Isoform) β-Arrestin Cell Line, 2 vials. (93-0351C2)
Supplier:  Eurofins Discoverx 930351C2
Encompass
The PathHunter DRD2S cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine) based activation of DRD2S GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged DRD2S and EA-tagged β-Arrestin-2. Activation of DRD2S stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0328
Add to Cart
Request a Quote