Learn More
Eurofins Discoverx PathHunter CHO-K1 GPR120S (Short Isoform) β-Arrestin-1 Cell Line, 2 vials. (93-0978C2)
Catalog No. 502001728
Encompass
The PathHunter GPR120S (ARR1) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of GPR120S GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR120S and EA-tagged β-Arrestin-1. Activation of GPR120S stimulates the recruitment of β-Arrestin-1 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1728 Supplier Eurofins Discoverx Supplier No. 930978C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter GPR120S (ARR1) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of GPR120S GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR120S and EA-tagged β-Arrestin-1. Activation of GPR120S stimulates the recruitment of β-Arrestin-1 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.