Learn More
Molecular Devices Molecular Devices CHO Growth A Medium, Semi-solid, L-Gln, 90 mL
Catalog No. 5000014449
Encompass_Preferred
Semi-solid cloning medium formulated for suspension-adapted Chinese hamster ovary (CHO) cells. Contains L-glutamine and methylcellulose to immobilize single cells and promote discrete colony formation. Supplied as a single 90 mL ready-to-use aliquot for research use.
  • Semi-solid medium containing methylcellulose and L-glutamine.
  • Supports CHO, DG44, and DUXB11 suspension cell cloning.
  • Immobilizes single cells for discrete colony formation.
  • Single 90 mL ready-to-use aliquot for plating or automated workflows.
  • No material of animal origin used in manufacture.
  • For research use only; not for clinical use.
Catalog No. 50-000-14449 Supplier Molecular Devices Supplier No. K8810
Semi-solid cloning medium formulated for suspension-adapted Chinese hamster ovary (CHO) cells. Contains L-glutamine and methylcellulose to immobilize single cells and promote discrete colony formation. Supplied as a single 90 mL ready-to-use aliquot for research use.
  • Semi-solid medium containing methylcellulose and L-glutamine.
  • Supports CHO, DG44, and DUXB11 suspension cell cloning.
  • Immobilizes single cells for discrete colony formation.
  • Single 90 mL ready-to-use aliquot for plating or automated workflows.
  • No material of animal origin used in manufacture.
  • For research use only; not for clinical use.